Merck
Gd
Physical chemistry chemical physics : PCCP (2017-02-01)
Jessica A Clayton, Mian Qi, Adelheid Godt, Daniella Goldfarb, Songi Han, Mark S Sherwin
ABSTRACT
Electron paramagnetic resonance spectroscopy in combination with site-directed spin labeling is a very powerful tool for elucidating the structure and organization of biomolecules. Gd