- Ru(II) carbazole thiosemicarbazone complexes with four membered chelate ring: Synthesis, molecular structures and evaluation of biological activities.
Ru(II) carbazole thiosemicarbazone complexes with four membered chelate ring: Synthesis, molecular structures and evaluation of biological activities.
Journal of photochemistry and photobiology. B, Biology (2016-11-14)
Ramasamy Raj Kumar, Rengan Ramesh, Jan Grzegorz Małecki
PMID27838485
ABSTRACT
Formation of ruthenium(II) complexes of the type [RuH(CO)(PPh