Merck
  • Bcl-3 regulates TGFβ signaling by stabilizing Smad3 during breast cancer pulmonary metastasis.
Bcl-3 regulates TGFβ signaling by stabilizing Smad3 during breast cancer pulmonary metastasis.
Cell death & disease (2016-12-03)
Xi Chen, Xinwei Cao, Xiaohua Sun, Rong Lei, Pengfei Chen, Yongxu Zhao, Yuhang Jiang, Jie Yin, Ran Chen, Deji Ye, Qi Wang, Zhanjie Liu, Sanhong Liu, Chunyan Cheng, Jie Mao, Yingyong Hou, Mingliang Wang, Ulrich Siebenlist, Y Eugene Chin, Ying Wang, Liu Cao, Guohong Hu, Xiaoren Zhang
ABSTRACT
Transforming growth factor beta (TGFβ) signaling in breast cancer is selectively associated with pulmonary metastasis. However, the underlying mechanisms remain unclear. Here we show that Bcl-3, a member of the IκB family, serves as a critical regulator in TGFβ signaling to modulate breast cancer pulmonary metastasis. Bcl-3 expression was significantly associated with metastasis-free survival in breast cancer patients. Bcl-3 deletion inhibited the migration and invasion of breast cancer cells in vitro, as well as breast cancer lung metastasis in vivo. Bcl-3 was required for the expression of downstream TGFβ signaling genes that are involved in breast cancer lung metastasis. Bcl-3 knockdown enhanced the degradation of Smad3 but not Smad2 following TGFβ treatment. Bcl-3 could bind to Smad3 and prevent the ubiquitination and degradation of Smad3 protein. These results indicate that Bcl-3 serves as a promising target to prevent breast tumor lung metastasis.