- Excitonic Splitting and Vibronic Coupling Analysis of the m-Cyanophenol Dimer.
Excitonic Splitting and Vibronic Coupling Analysis of the m-Cyanophenol Dimer.
The journal of physical chemistry. A (2016-12-22)
Franziska A Balmer, Sabine Kopec, Horst Köppel, Samuel Leutwyler
PMID28001398
ABSTRACT
The S