Merck
  • Lipid Class Specific Quantitative Analysis of n-3 Polyunsaturated Fatty Acids in Food Supplements.
Lipid Class Specific Quantitative Analysis of n-3 Polyunsaturated Fatty Acids in Food Supplements.
Journal of agricultural and food chemistry (2016-12-23)
Laura Kutzner, Annika I Ostermann, Thade Konrad, Dieter Riegel, Stefan Hellhake, Jan Philipp Schuchardt, Nils Helge Schebb
ABSTRACT
Supplementation products containing n-3 PUFA from marine sources serve a large market. Although the amount of eicosapentaenoic acid and docosahexaenoic acid in the products is provided by the manufacturer, no or little information is available on their lipid pattern. Therefore, we quantitatively analyzed the fatty acid pattern in the lipid fractions triglycerides, phospholipids, ethyl esters, and free fatty acids in supplementation products by means of solid phase extraction and gas chromatography. Twelve products from the European and U.S. markets containing fish, krill, algal, or plant oil were analyzed. Total n-3 PUFA content ranged from 68 g/100 g fat (fish oil) to 42 g/100 g fat (algal oil) to 17 g/100 g fat (krill oil). On the basis of the n-3 PUFA containing lipid class, the supplements can be separated dominantly in ethyl ester, re-esterified triglyceride, triglyceride, and phospholipid containing products. Algae-based products contained natural triglycerides, krill oils a complex mixture of phospholipids, triglycerides, and free fatty acids, and fish oil products either ethyl esters, re-esterified triglycerides, or triglycerides. Even products of the same class and source showed distinct differences in their lipid pattern. A specification of the lipid composition of n-3 PUFA products would allow distinguishing the different (qualities of) supplements.