- Estradiol stimulates glycogen synthesis whereas progesterone promotes glycogen catabolism in the uterus of the American mink (Neovison vison).
Estradiol stimulates glycogen synthesis whereas progesterone promotes glycogen catabolism in the uterus of the American mink (Neovison vison).
Animal science journal = Nihon chikusan Gakkaiho (2016-05-14)
Kole Bowman, Jack Rose
PMID27170562
ABSTRACT
Glycogen synthesis by mink uterine glandular and luminal epithelia (GE and LE) is stimulated by estradiol (E