Merck
  • Continuous microwave pasteurization of a vegetable smoothie improves its physical quality and hinders detrimental enzyme activity.
Continuous microwave pasteurization of a vegetable smoothie improves its physical quality and hinders detrimental enzyme activity.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional (2016-06-28)
Mitra Arjmandi, Mariano Otón, Francisco Artés, Francisco Artés-Hernández, Perla A Gómez, Encarna Aguayo
ABSTRACT
The effect of a pasteurization treatment at 90 ± 2 ℃ for 35 s provided by continuous microwave under different doses (low power/long time and high power/short time) or conventional pasteurization on the quality of orange-colored smoothies and their changes throughout 45 days of storage at 5 ℃ was investigated. A better color retention of the microwave pasteurization- treated smoothie using high power/short time than in conventionally processed sample was evidenced by the stability of the hue angle. The continuous microwave heating increased the viscosity of the smoothie more than the conventional pasteurization in comparison with non-treated samples. Lower residual enzyme activities from peroxidase, pectin methylesterase and polygalacturonase were obtained under microwave heating, specifically due to the use of higher power/shorter time. For this kind of smoothie, polygalacturonase was the more thermo-resistant enzyme and could be used as an indicator of pasteurization efficiency. The use of a continuous semi-industrial microwave using higher power and shorter time, such as 1600 W/206 s and 3600 W/93 s, resulted in better quality smoothies and greater enzyme reduction than conventional thermal treatment.