Merck
  • NKp30+ NK cells are associated with HBV control during pegylated-interferon-alpha-2b therapy of chronic hepatitis B.
NKp30+ NK cells are associated with HBV control during pegylated-interferon-alpha-2b therapy of chronic hepatitis B.
Scientific reports (2016-12-13)
Xiaokun Shen, Binqing Fu, Yanyan Liu, Chuang Guo, Ying Ye, Rui Sun, Jiabin Li, Zhigang Tian, Haiming Wei
ABSTRACT
A pressing need exists for improved therapeutic options for chronic hepatitis B (CHB). Pegylated-interferon-alpha (Peg-IFN-α) achieves sustained off-treatment responses in many cases because of its direct anti-viral effects and regulation of the immune response. However, non-responsiveness to Peg-IFN-α is frequent, and the mechanism is poorly understood. In this study, we found that the frequency and absolute number of NKp30+ natural killer (NK) cells increased markedly, accompanied by enhanced CD107a and IFN-γ production, during Peg-IFN-α-2b monotherapy or combination therapy with adefovir dipivoxil in patients with CHB, especially in responders. The responders and non-responders differed in the frequency of polyfunctional IFN-γ+ CD107+ NK cells. In addition, the increase in NKp30+ NK cells was negatively correlated with the HBV viral load and plasma HBeAg. Moreover, it was found that IL-15 may contribute to the up-regulation of NKp30 on the NK cells, and this up-regulation was not induced in vitro by Peg-IFN-α-2b alone. However, in the non-responders, these NKp30+ NK cells were dysfunctional because of increased NKG2A expression, which partly explains the inactivation of NKp30+ NK cells and the reduced capacity of these cells to produce antiviral cytokines. These findings may provide a new mechanism to explain the variable efficacy of Peg-IFN-α-2b therapy.