Merck
  • In vivo production of interferon beta by human Tenon's fibroblasts; a possible mediator for the development of chronic conjunctival inflammation.
In vivo production of interferon beta by human Tenon's fibroblasts; a possible mediator for the development of chronic conjunctival inflammation.
The British journal of ophthalmology (2002-05-30)
L Chang, D Siriwardena, M R Wilkins, J G Crowston, A N Akbar, P T Khaw
ABSTRACT
Chronic inflammation may develop from failure of the immune system to deactivate itself during resolution of the wound healing response, and is recognised as a major risk factor for trabeculectomy failure. Fibroblast/T cell interactions may contribute to aggressive scarring. Our previous research showed that in vitro human Tenon's fibroblast produced interferon beta was responsible for preventing T cell apoptosis, suggesting that this interaction could contribute to the development of chronic inflammation. Immunohistological techniques were used to investigate the in vivo components of this particular fibroblast/T cell interaction in conjunctival biopsies from glaucoma patients undergoing filtration surgery. Fibroblast produced interferon beta and T lymphocytes were identified in human conjunctiva. The components of fibroblast mediated prevention of T cell apoptosis were identified in vivo, suggesting that the development of this interaction is possible and that it may contribute to the development of chronic inflammation and excessive scarring.