Merck
  • Evaluation of Different DNA Vaccines against Porcine Reproductive and Respiratory Syndrome (PRRS) in Pigs.
Evaluation of Different DNA Vaccines against Porcine Reproductive and Respiratory Syndrome (PRRS) in Pigs.
Vaccines (2013-01-01)
Stefano Petrini, Giorgio Ramadori, Riccardo Villa, Paolo Borghetti, Elena de Angelis, Anna Maria Cantoni, Attilio Corradi, Augusto Amici, Maura Ferrari
ABSTRACT
In veterinary medicine, there have been different experiences with the plasmid DNA vaccination. In this area and with the hypothesis to demonstrate the effectiveness of different plasmids encoding porcine respiratory and reproductive syndrome (PRRS), five DNA vaccines against PRRS were evaluated for their innocuity and efficacy in pigs. Eighteen animals were divided into five groups which were injected with five (A, B, C, D, E) different DNA vaccines. Albeit, none of the proposed vaccines were able to protect the animals against PRRS virus. Only vaccines A and B were able to reduce the clinical signs of the infection. ELISA IgM were detected 30 days after the first vaccination in the pigs injected by Vaccine A or B. ELISA IgG were detected 90 days after the first vaccination in the pigs injected by Vaccine B or C. Neutralizing antibody were detected Post Challenge Days 61 (PCD) in all groups. In the pigs inoculated with Vaccine C, IFN-g were detected 90 days after first vaccination, and after challenge exposure they increased. In the other groups, the IFN-g were detected after challenge infection. Pigs injected with each of the vaccines A, B, C, D and E showed a significantly higher level of CD4(-)CD8⁺ lymphocytes (p < 0.001) after infection in comparison with their controls.