- Potentials and capabilities of the Extracellular Vesicle (EV) Array.
Potentials and capabilities of the Extracellular Vesicle (EV) Array.
Journal of extracellular vesicles (2015-04-12)
Malene Møller Jørgensen, Rikke Bæk, Kim Varming
PMID25862471
ABSTRACT
Extracellular vesicles (EVs) and exosomes are difficult to enrich or purify from biofluids, hence quantification and phenotyping of these are tedious and inaccurate. The multiplexed, highly sensitive and high-throughput platform of the EV Array presented by Jørgensen et al., (J Extracell Vesicles, 2013; 2: 10) has been refined regarding the capabilities of the method for characterization and molecular profiling of EV surface markers. Here, we present an extended microarray platform to detect and phenotype plasma-derived EVs (optimized for exosomes) for up to 60 antigens without any enrichment or purification prior to analysis.