Merck
  • Immunohistochemical expression of ARID1A in penile squamous cell carcinomas: a tissue microarray study of 112 cases.
Immunohistochemical expression of ARID1A in penile squamous cell carcinomas: a tissue microarray study of 112 cases.
Human pathology (2015-03-18)
Sheila F Faraj, Alcides Chaux, Nilda Gonzalez-Roibon, Enrico Munari, Antonio L Cubilla, Ie-Ming Shih, George J Netto
ABSTRACT
ARID1A, a member of the chromatin remodeling genes family, has been suggested as a novel tumor suppressor gene in gynecologic malignancies. However, its role in penile cancer has yet to be determined. This study assesses the immunohistochemical expression of ARID1A in penile squamous cell carcinoma (SCC) and its association with pathologic features, human papillomavirus (HPV) status, and previously reported mammalian target of rapamycin pathway markers in the same cohort. Four tissue microarrays were constructed from 112 cases of formalin-fixed, paraffin-embedded penile SCC from Paraguay. Each tumor was sampled 3 to 12 times. ARID1A expression was evaluated by immunohistochemistry using a polyclonal rabbit anti-ARID1A (BAF250A) antibody. An H score was calculated in each spot as the sum of expression intensity (0-3+) by extent (0%-100%). Median H score per case was used for statistical analysis. ARID1A expression was observed in all cases, ranging from 3% to 100% of tumor cells (median, 95%). In 96 cases (86%), ARID1A expression was observed in 90% or more tumor cells. HPV DNA was detected in 20 (38%) of 52 analyzed samples. There was a significant trend of association between ARID1A and histologic grade. ARID1A expression was not associated with histologic subtype (P = .61) or HPV status (P = .18). ARID1A expression decreased with decreasing levels of PTEN expression (P = .01). ARID1A was expressed in penile SCC, in most cases at high levels. A significant trend of association was found between histologic grade and ARID1A expression, with lower ARID1A expression, lower histologic grades, and decreased PTEN expression.