- The alarmin IL-33 is a notch target in quiescent endothelial cells.
The alarmin IL-33 is a notch target in quiescent endothelial cells.
The molecular mechanisms that drive expression of the alarmin interleukin-33 (IL-33) in endothelial cells are unknown. Because nuclear IL-33 is a marker of endothelial cell quiescence (corroborated in this study by coexpression of cyclin-dependent kinase inhibitor p27(Kip1)), we hypothesized that Notch signaling might be involved in regulating IL-33 expression. Activation of Notch1 by immobilized Notch ligands was sufficient to induce nuclear IL-33 expression in cultured endothelial cells. Conversely, IL-33 expression was inhibited by the γ-secretase inhibitor DAPT or by inhibiting the function of Dll4, Jagged1, Notch1, or the canonical Notch transcription factor RBP-Jκ. Insensitivity to cycloheximide indicated that IL-33 was a direct target of Notch signaling, well in line with the identification of several conserved RBP-Jκ binding sites in the IL33 gene. The in vivo expression of Dll4 but not of Jagged1 was well correlated with expression of IL-33 in quiescent vessels, and subcutaneous injection of DAPT in healthy skin reduced IL-33 expression, indicating that Notch signaling was involved. On the other hand, loss of IL-33 during angiogenesis occurred despite sustained Dll4 and Notch1 expression, suggesting that other signals may override the IL-33-driving signal in this context. Taken together, our data demonstrate that endothelial nuclear IL-33 is induced by Notch and that Dll4 may be the dominant ligand responsible for this signaling in vivo.