Merck
  • Synthesis of a ricin toxin B subunit-rotavirus VP7 fusion protein in potato.
Synthesis of a ricin toxin B subunit-rotavirus VP7 fusion protein in potato.
Molecular biotechnology (2006-01-31)
Nak-Won Choi, Mary K Estes, William H R Langridge
ABSTRACT
A gene encoding the outer capsid glycoprotein (VP7) of simian rotavirus SA11, was genetically linked to the amino terminus of the ricin toxin B subunit (RTB) isolated from castor-oil plant (Ricinus communis) seeds. To assess fusion protein expression in plant cells, the VP7::RTB fusion gene was transferred into potato (Solanum tuberosum) cells by Agrobacterium tumefaciens-mediated transformation methods and transformed plants regenerated. The fusion gene was detected in transformed potato genomic DNA by polymerase chain reaction DNA amplification methods. Immunoblot analysis with anti-SA11 antiserum as the primary antibody verified the presence of VP7::RTB fusion protein in transformed potato tuber tissues. The plant-synthesized fusion protein bound RTB membrane receptors as measured by asialofetuin-enzyme-linked immunosorbent assay (ELISA). The ELISA results indicated that the VP7::RTB fusion protein was biologically active and made up approx 0.03% of total soluble transformed tuber protein. The biosynthesis of receptor binding VP7::RTB fusion protein in potato tissues demonstrates the feasibility of producing monomeric ricin toxin B subunit adjuvant-virus antigen fusion proteins in crop plants for enhanced immunity.