Merck
  • PK/TD modeling for prediction of the effects of 8C2, an anti-topotecan mAb, on topotecan-induced toxicity in mice.
PK/TD modeling for prediction of the effects of 8C2, an anti-topotecan mAb, on topotecan-induced toxicity in mice.
International journal of pharmaceutics (2014-02-11)
Dhaval K Shah, Joseph P Balthasar
ABSTRACT
To facilitate the development of an inverse targeting strategy, where anti-topotecan antibodies are administered to prevent systemic toxicity following intraperitoneal topotecan, a pharmacokinetic/toxicodynamic (PK/TD) model was developed and evaluated. The pharmacokinetics of 8C2, a monoclonal anti-topotecan antibody, were assessed following IV and SC administration, and the data were characterized using a two compartmental model with nonlinear absorption and elimination. A hybrid PK model was constructed by combining a PBPK model for topotecan with the two-compartment model for 8C2, and the model was employed to predict the disposition of topotecan, 8C2, and the topotecan-8C2 complex. The model was linked to a toxicodynamic model for topotecan-induced weight-loss, and simulations were conducted to predict the effects of 8C2 on the toxicity of topotecan in mice. Increasing the molar dose ratio of 8C2 to topotecan resulted in a dose-dependent decrease in the unbound (i.e., not bound to 8C2) topotecan exposure in plasma (AUCf) and a decrease in the extent of topotecan-induced weight-loss. Consistent with model predictions, toxicodynamic experiments showed substantial reduction in the percent nadir weight loss observed with 30 mg/kg IP topotecan after co-administration of 8C2 (20 ± 8% vs. 10 ± 8%). The investigation supports the use of anti-topotecan mAb to reduce the systemic toxicity of IP topotecan chemotherapy.