Merck
  • Localization of Kv1.3 channels in the immunological synapse modulates the calcium response to antigen stimulation in T lymphocytes.
Localization of Kv1.3 channels in the immunological synapse modulates the calcium response to antigen stimulation in T lymphocytes.
Journal of immunology (Baltimore, Md. : 1950) (2009-10-21)
Stella A Nicolaou, Lisa Neumeier, Ashleigh Steckly, Vladimir Kucher, Koichi Takimoto, Laura Conforti
ABSTRACT
The immunological synapse (IS), a highly organized structure that forms at the point of contact between a T cell and an APC, is essential for the proper development of signaling events, including the Ca(2+) response. Kv1.3 channels control Ca(2+) homeostasis in human T cells and move into the IS upon Ag presentation. However, the process involved in channel accumulation in the IS and the functional implications of this localization are not yet known. Here we define the movement of Kv1.3 into the IS and study whether Kv1.3 localization into the IS influences Ca(2+) signaling in Jurkat T cells. Crosslinking of the channel protein with an extracellular Ab limits Kv1.3 mobility and accumulation at the IS. Moreover, Kv1.3 recruitment to the IS does not involve the transport of newly synthesized channels and it does not occur through recycling of membrane channels. Kv1.3 localization in the IS modulates the Ca(2+) response. Blockade of Kv1.3 movement into the IS by crosslinking significantly increases the amplitude of the Ca(2+) response triggered by anti-CD3/anti-CD28-coated beads, which induce the formation of the IS. On the contrary, the Ca(2+) response induced by TCR stimulation without the formation of the IS with soluble anti-CD3/anti-CD28 Abs is unaltered. The results presented herein indicate that, upon Ag presentation, membrane-incorporated Kv1.3 channels move along the plasma membrane to localize in the IS. This localization is important to control the amplitude of the Ca(2+) response, and disruption of this process can account for alterations of downstream Ca(2+)-dependent signaling events.