- Msi RNA-binding proteins control reserve intestinal stem cell quiescence.
Msi RNA-binding proteins control reserve intestinal stem cell quiescence.
The Journal of cell biology (2016-11-02)
Maryam Yousefi, Ning Li, Angela Nakauka-Ddamba, Shan Wang, Kimberly Davidow, Jenna Schoenberger, Zhengquan Yu, Shane T Jensen, Michael G Kharas, Christopher J Lengner
PMID27799368
ABSTRACT
Regeneration of the intestinal epithelium is driven by multiple intestinal stem cell (ISC) types, including an active, radiosensitive Wnt