- Achieving Fully Reversible Conversion in MoO
Achieving Fully Reversible Conversion in MoO
ACS nano (2016-11-05)
Wei Wang, Jinwen Qin, Zhigang Yin, Minhua Cao
PMID27809481
ABSTRACT
Electrode materials based on conversion reactions with lithium ions generally show much higher energy density. One of the main challenges in the design of these electrode materials is to improve initial Coulombic efficiency and alleviate the volume changes during the lithiation-delithiation processes. Here, we achieve fully reversible conversion in MoO