- Ionic liquid induced G-quadruplex formation and stabilization: spectroscopic and simulation studies.
Ionic liquid induced G-quadruplex formation and stabilization: spectroscopic and simulation studies.
Physical chemistry chemical physics : PCCP (2016-11-03)
Sagar Satpathi, Mandar Kulkarni, Arnab Mukherjee, Partha Hazra
PMID27766324
ABSTRACT
Among different polymorphs of DNA, G-quadruplex (GQ) formation in guanine rich sequences has received special attention due to its direct relevance to cellular aging and abnormal cell growths. To date, smaller ions like Na