Merck
  • Psoriasis is associated with increased beta-defensin genomic copy number.
Psoriasis is associated with increased beta-defensin genomic copy number.
Nature genetics (2007-12-07)
Edward J Hollox, Ulrike Huffmeier, Patrick L J M Zeeuwen, Raquel Palla, Jesús Lascorz, Diana Rodijk-Olthuis, Peter C M van de Kerkhof, Heiko Traupe, Gys de Jongh, Martin den Heijer, André Reis, John A L Armour, Joost Schalkwijk
ABSTRACT
Psoriasis is a common inflammatory skin disease with a strong genetic component. We analyzed the genomic copy number polymorphism of the beta-defensin region on human chromosome 8 in 179 Dutch individuals with psoriasis and 272 controls and in 319 German individuals with psoriasis and 305 controls. Comparisons in both cohorts showed a significant association between higher genomic copy number for beta-defensin genes and risk of psoriasis.