Merck
  • ROS-mediated Different Homeostasis of Murine Corneal Epithelial Progenitor Cell Line under Oxidative Stress.
ROS-mediated Different Homeostasis of Murine Corneal Epithelial Progenitor Cell Line under Oxidative Stress.
Scientific reports (2016-11-03)
Jing Zhou, Lianping Ge, Changkai Jia, Xiling Zheng, Huixia Cui, Rongrong Zong, Xiaorui Bao, Yuanyuan Yin, Jian-Xing Ma, Wei Li, Zuguo Liu, Yueping Zhou
ABSTRACT
The role of ROS in stem cell biology has not been fully illustrated and understood. Here we compared the different responses and investigated the mechanism underlying oxidative stress induced by hydrogen peroxide (H2O2) between murine corneal epithelial progenitor cell line (TKE2) and mature murine corneal epithelial cells (MCE). TKE2 showed a different homeostasis and strong resistance to H2O2. TKE2 reduced the production of ROS, inhibited ROS generation enzyme NADPH oxidase 4 (NOX4), and increased dual specificity phosphatase 6 (DUSP6). Furthermore, TKE2 activated nuclear factor (erythroid-derived 2)-like 2 (NRF2) signaling pathway, regulated miR-125B1 and miR-29B1, and elevated levels of antioxidants glutathione S-transferase P (GSTP) and superoxide dismutases (SOD). The association with ROS of the cells was also verified by RNA interference approach and pharmacological antagonization. In addition, TKE2 enhanced the autophagy after exposure to H2O2. The novel evidence suggests that TKE2 cells have different homeostasis and strong antioxidant properties against oxidative stress via the regulation of ROS formation and pathway.
Sigma-Aldrich
Anti-BCRP Antibody, clone BXP-21, culture supernatant, clone BXP-21, Chemicon®