- Surface-bound bovine serum albumin carrier protein as present in recombinant cytokine preparations amplifies T helper 17 cell polarization.
Surface-bound bovine serum albumin carrier protein as present in recombinant cytokine preparations amplifies T helper 17 cell polarization.
Scientific reports (2016-11-04)
Lei Dong, Alexandra Helmke, Ari Waisman, Hermann Haller, Andreas Pich, Sibylle von Vietinghoff
PMID27808281
ABSTRACT
Understanding of T helper 17 lineage (T