Merck
  • Inhibition of integrin-mediated cell adhesion but not directional cell migration requires catalytic activity of EphB3 receptor tyrosine kinase. Role of Rho family small GTPases.
Inhibition of integrin-mediated cell adhesion but not directional cell migration requires catalytic activity of EphB3 receptor tyrosine kinase. Role of Rho family small GTPases.
The Journal of biological chemistry (2004-11-13)
Hui Miao, Klaus Strebhardt, Elena B Pasquale, Tang-Long Shen, Jun-Lin Guan, Bingcheng Wang
ABSTRACT
Genetic studies have shown that Eph receptor tyrosine kinases have both kinase-dependent and kinase-independent functions through incompletely understood mechanisms. We report here that ephrin-B1 stimulation of endogenous EphB kinases in LS174T colorectal epithelial cells inhibited integrin-mediated adhesion and HGF/SF-induced directional cell migration. Using 293 cells stably transfected with wild type (WT)- or kinase-deficient (KD-EphB3), we found that inhibition of integrin-mediated cell adhesion and induction of cell rounding was kinase-dependent. Unexpectedly, in two independent assays, both KD- and WT-EphB3 significantly inhibited directional cell migration. Upon ephrin-B1 stimulation, the activities of Rac1 and Cdc42 were reduced in both WT- and KD-EphB3-expressing cells that were induced to migrate. Pharmacological evidence demonstrates that a relative increase in RhoA signaling as a result of decreased Rac1/Cdc42 activities contributes to the inhibitory effects. Furthermore, EphB3-mediated inhibitory effect on cell adhesion but not migration was abolished by the integrin activating antibodies, suggesting that the inhibition of cell migration is not because of down-regulation of integrin function. These results uncover a differential requirement for EphB3 catalytic activity in the regulation of cell adhesion and migration, and suggest that while catalytic activity of EphB3 is required for inhibition of integrin-mediated cell adhesion, a distinct signaling pathway to Rho GTPases shared by WT- and KD-EphB3 receptor mediates inhibition of directional cell migration.