Merck
  • Development of novel inhibitor probes of DNA polymerase III based on dGTP analogs of the HPUra type: base, nucleoside and nucleotide derivatives of N2-(3,4-dichlorobenzyl)guanine.
Development of novel inhibitor probes of DNA polymerase III based on dGTP analogs of the HPUra type: base, nucleoside and nucleotide derivatives of N2-(3,4-dichlorobenzyl)guanine.
Nucleic acids research (1990-12-25)
M M Butler, L W Dudycz, N N Khan, G E Wright, N C Brown
ABSTRACT
6-(p-Hydroxyphenylhydrazino)uracil (H2-HPUra) is a selective and potent inhibitor of the replication-specific class III DNA polymerase (pol III) of Gr+ bacteria. Although formally a pyrimidine, H2-HPUra derives its inhibitory activity from its specific capacity to mimic the purine nucleotide, dGTP. We describe the successful conversion of the H2-HPUra inhibitor prototype to a bona fide purine, using N2-(benzyl)guanine (BG) as the basis. Structure-activity relationships of BGs carrying a variety of substituents on the aryl ring identified N2-(3,4-dichlorobenzyl)guanine (DCBG) as a nucleus equivalent to H2-HPUra with respect to potency and inhibitor mechanism. DCBdGTP, the 2'-deoxyribonucleoside 5'-triphosphate form of DCBG, was synthesized and characterized with respect to its action on wild-type and mutant forms of B. subtilis DNA pol III. DCBdGTP acted on pol III by the characteristic inhibitor mechanism and formally occupied the dNTP binding site with a fit which permitted its polymerization.