Merck
  • Novel sulfamide analogs of oleoylethanolamide showing in vivo satiety inducing actions and PPARalpha activation.
Novel sulfamide analogs of oleoylethanolamide showing in vivo satiety inducing actions and PPARalpha activation.
Journal of medicinal chemistry (2007-01-19)
Carolina Cano, Javier Pavón, Antonia Serrano, Pilar Goya, Juan Antonio Paez, Fernando Rodriguez de Fonseca, Manuel Macias-Gonzalez
ABSTRACT
Long chain saturated and unsaturated alkyl sulfamide and propyl sulfamide derivatives, analogs of oleoylethanolamide, have been synthesized and evaluated in vivo and in vitro as peroxisome proliferator activated receptor alpha (PPARalpha) activators. Additionally, the anorexic effects of the new compounds have been studied in vivo in food-deprived rats. Among the active compounds N-octadecyl-N'-propylsulfamide (7) has been identified as a potent hypolipidemic compound, a potent feeding suppressant, and a concentration-dependent activator of PPARalpha.