Merck
  • Screening study of SFC critical method parameters for the determination of pharmaceutical compounds.
Screening study of SFC critical method parameters for the determination of pharmaceutical compounds.
Journal of pharmaceutical and biomedical analysis (2016-04-23)
Amandine Dispas, Pierre Lebrun, Pierre-Yves Sacré, Philippe Hubert
ABSTRACT
Nowadays, supercritical fluid chromatography is commonly presented as a promising alternative technique in the field of separation sciences. Nevertheless the selection of chromatographic conditions and sample preparation of pharmaceutical compounds remain a challenge and peak distortion was previously highlighted. The main objective of the present work was to evaluate the impact of different critical method parameters (CMPs), i.e. stationary phase, mobile phase composition and injection solvent nature. The experiments were performed considering two groups of antimalarial molecules: one group with neutral/apolar compounds and the other one with salt form of polar compounds. In this context, another objective was to propose a suitable sample solvent for quantitative analysis. The interest of new generation stationary phase to obtain good peak shape and the interest to tune the mobile phase composition were demonstrated. During this study, design of experiments and desirability function approach enabled to highlight optimal chromatographic conditions in order to maximise peak capacity and to get acceptable value of symmetry factor. Regarding sample injection solvent composition, some counterintuitive results were observed: solvents closer to the mobile phase polarity (i.e heptane or 2-propanol/heptane mixture) did not provide best results in terms of peak symmetry. In addition, acetonitrile and short aliphatic alcohols offered an interesting alternative as injection solvent: toxicity of solvents used is clearly reduced and better quantitative performances could be expected while keeping high peak capacity and symmetric sharp peaks. Finally, the quantitative performances were evaluated by the method validation for the quantitative determination of quinine sulfate in a pharmaceutical formulation. These better understandings on critical method parameters led SFC to be an even more promising technique in the field of the analysis of pharmaceutical compounds.