Merck
  • Study of breast cancer incidence in patients of lymphangioleiomyomatosis.
Study of breast cancer incidence in patients of lymphangioleiomyomatosis.
Breast cancer research and treatment (2016-03-10)
Olivier Nuñez, Antonio Román, Simon R Johnson, Yoshikazu Inoue, Masaki Hirose, Álvaro Casanova, Gorka Ruiz de Garibay, Carmen Herranz, Gema Bueno-Moreno, Jacopo Boni, Francesca Mateo, Anna Petit, Fina Climent, Teresa Soler, August Vidal, José Vicente Sánchez-Mut, Manel Esteller, José Ignacio López, Nadia García, Anna Gumà, Raúl Ortega, María Jesús Plà, Miriam Campos, Emilio Ansótegui, María Molina-Molina, Claudia Valenzuela, Piedad Ussetti, Rosalía Laporta, Julio Ancochea, Antoni Xaubet, Marina Pollán, Miguel Angel Pujana
ABSTRACT
Molecular evidence has linked the pathophysiology of lymphangioleiomyomatosis (LAM) to that of metastatic breast cancer. Following on this observation, we assessed the association between LAM and subsequent breast cancer. An epidemiological study was carried out using three LAM country cohorts, from Japan, Spain, and the United Kingdom. The number of incident breast cancer cases observed in these cohorts was compared with the number expected on the basis of the country-specific incidence rates for the period 2000-2014. Immunohistochemical studies and exome sequence analysis were performed in two and one tumors, respectively. All cohorts revealed breast cancer standardized incidence ratios (SIRs) ≥ 2.25. The combined analysis of all cases or restricted to pre-menopausal age groups revealed significantly higher incidence of breast cancer: SIR = 2.81, 95 % confidence interval (CI) = 1.32-5.57, P = 0.009; and SIR = 4.88, 95 % CI = 2.29-9.99, P = 0.0007, respectively. Immunohistochemical analyses showed positivity for known markers of lung metastatic potential. This study suggests the existence of increased breast cancer risk among LAM patients. Prospective studies may be warranted to corroborate this result, which may be particularly relevant for pre-menopausal women with LAM.
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit