Merck
  • Determination of Vitamin E and Vitamin A in Infant Formula and Adult Nutritionals by Normal-Phase High-Performance Liquid Chromatography: Collaborative Study, Final Action 2012.10.
Determination of Vitamin E and Vitamin A in Infant Formula and Adult Nutritionals by Normal-Phase High-Performance Liquid Chromatography: Collaborative Study, Final Action 2012.10.
Journal of AOAC International (2016-02-10)
Adrienne McMahon
ABSTRACT
The main objective of the AOAC Stakeholder Panel on Infant Formula and Adult Nutritionals (SPIFAN) project is to establish international consensus methods for infant formula and adult nutritionals, which will benefit intermarket supply and dispute resolution. A collaborative study was conducted on AOAC First Action Method 2012.10 Simultaneous Determination of 13-cis and All-trans Vitamin A Palmitate (Retinyl Palmitate), Vitamin A Acetate (Retinyl Acetate), and Total Vitamin E (α-Tocopherol and D-α-tocopherol acetate) in Infant Formula and Adult Nutritionals by Normal-Phase HPLC. Fifteen laboratories from 11 countries participated in an interlaboratory study to determine 13-cis and all-trans vitamin A palmitate (retinyl palmitate), vitamin A acetate (retinyl acetate), and total vitamin E (α-tocopherol and D-α-tocopherol acetate) in infant formula and adult nutritionals by normal-phase HPLC and all laboratories returned valid data. Eighteen test portions of nine blind duplicates of a variety of infant formula and adult nutritional products were used in the study. The matrixes included milk-based and soy-based hydrolyzed protein as well as a low fat product. Each of the samples was prepared fresh and analyzed in singlicate. As the number of samples exceeded the recommended number to be prepared in a single day, analysis took place over 2 days running 12 samples on day one and 10 samples on day two. The reference standard stock was prepared once and the six-point curve diluted freshly on each day. Results obtained from all 15 laboratories are reported. The RSDR for total vitamin A (palmitate or acetate) ranged from 6.51 to 22.61% and HorRat values ranged from 0.33 to 1.25. The RSDR for total vitamin E (as tocopherol equivalents) ranged from 3.84 to 10.78% and HorRat values ranged from 0.27 to 1.04. Except for an adult low fat matrix which generated reproducibility RSD >40% for some isomers, most SPIFAN matrixes gave results within the acceptance criteria of <16% RSD as stated in the respective Standard Method Performance Requirements.
Sigma-Aldrich
Papain from Carica papaya, powder, ≥3 U/mg