Merck
  • Identification of BRAF as a new interactor of PLCepsilon1, the protein mutated in nephrotic syndrome type 3.
Identification of BRAF as a new interactor of PLCepsilon1, the protein mutated in nephrotic syndrome type 3.
American journal of physiology. Renal physiology (2007-10-19)
Hassan Chaib, Bethan E Hoskins, Shazia Ashraf, Meera Goyal, Roger C Wiggins, Friedhelm Hildebrandt
ABSTRACT
Steroid-resistant nephrotic syndrome is a malfunction of the kidney glomerular filter that leads to proteinuria, hypoalbuminemia, edema, and renal failure. Recently, we identified recessive mutations in the phospholipase C epsilon 1 gene (PLCE1) as a new cause of early-onset nephrotic syndrome and demonstrated interaction of PLCepsilon1 with IQGAP1. To further elucidate the mechanism by which PLCE1 mutations cause nephrotic syndrome, we sought to identify new protein interaction partners of PLCepsilon1. We utilized information from the genetic interaction network of C. elegans. It relates the PLCE1 ortholog (plc-1) to the C. elegans ortholog (lin-45) of human BRAF (v-raf murine sarcoma viral oncogene homolog B1). We hypothesized that this may indicate a functional protein-protein interaction. Using GST pull down of HEK293T cell lysates in vitro and coimmunoprecipation of mouse kidney lysates in vivo, we show that BRAF interacts with PLCepsilon1. By immunohistochemistry in rat kidney, we demonstrate that both proteins are coexpressed and colocalize in developing and mature glomerular podocytes, reporting for the first time the expression of BRAF in the glomerular podocyte.