- Gold-catalyzed synthesis of substituted tetrahydronaphthalenes.
Gold-catalyzed synthesis of substituted tetrahydronaphthalenes.
Organic letters (2006-12-01)
Christiane M Grisé, Louis Barriault
PMID17134302
ABSTRACT
We report a gold-catalyzed benzannulation of 3-hydroxy-1,5-enynes to generate tetrahydronaphthalenes. This mild process proves to be an effective method to synthesize various metasubstituted aromatic rings in good yields. [reaction: see text]