- Copper salt catalyzed addition of arylboronic acids to azodicarboxylates.
Copper salt catalyzed addition of arylboronic acids to azodicarboxylates.
The Journal of organic chemistry (2005-10-08)
Takeshi Uemura, Naoto Chatani
PMID16209627
ABSTRACT
The addition of arylboronic acids 1 to azodicarboxylates 2 in the presence of a catalytic amount of a copper salt under mild reaction conditions gives aryl-substituted hydrazines 3 in high yields. The reaction is tolerant of a wide variety of functional groups.