- Palladium nanoparticles entrapped in aluminum hydroxide: dual catalyst for alkene hydrogenation and aerobic alcohol oxidation.
Palladium nanoparticles entrapped in aluminum hydroxide: dual catalyst for alkene hydrogenation and aerobic alcohol oxidation.
Organic letters (2005-03-12)
Min Serk Kwon, Namdu Kim, Cheon Min Park, Jae Sung Lee, Kyung Yeon Kang, Jaiwook Park
PMID15760143
ABSTRACT
[reaction: see text] A new aluminum hydroxide-supported palladium catalyst (1) made by a one-pot synthesis through nanoparticle generation and gelation shows a dual catalytic activity for olefinic hydrogenation and aerobic alcohol oxidation.