- Metabonomic strategy to the evaluation of chinese medicine compound danshen dripping pills interfering myocardial ischemia in rats.
Metabonomic strategy to the evaluation of chinese medicine compound danshen dripping pills interfering myocardial ischemia in rats.
Coronary heart disease (CHD) is one of the highest mortality diseases in the world. Traditional Chinese medicine compound Danshen dripping pills (CDDPs) have currently made a great achievement in treating CHD. However, the therapeutic mechanism of CDDP is often poorly interpreted. In this study, a GC-MS-based metabonomic study was conducted to assess the holistic efficacy of CDDP for myocardial infarction in male Sprague-Dawley rats, which were divided into the control group, the sham group, the model group, the control + CDDP group, and the model + CDDP, with CDDP at a dose of 107 mg/kg·d (equal to 1.8 mL/kg·d). The metabonomic findings demonstrated great differences of metabolic pattern among sham, model, and the model + CDDP in the orthogonal partial least squares discriminant analysis (OPLS-DA) models, which coordinated well with the assessment of plasma biochemistry and histopathological assay. Differentially expressed metabolites suggested that energy metabolism, glycolysis, and lipid metabolism might be disrupted by myocardial infarction. Both the potential metabolic biomarkers and the biochemical histopathological indices were regulated effectively by CDDP.