Merck
  • Synthesis and Antimycobacterial Activity of 2-(Phenylthio) benzoylarylhydrazone Derivatives.
Synthesis and Antimycobacterial Activity of 2-(Phenylthio) benzoylarylhydrazone Derivatives.
Iranian journal of pharmaceutical research : IJPR (2011-10-01)
Ali Almasirad, Somayeh Samiee-Sadr, Abbas Shafiee
ABSTRACT
A series of new 2-(phenylthio) benzoylarylhydrazones were synthesized by acid-catalyzed condensation of hydrazide 3 with corresponding aldehydes. The chemical structures of the compounds were elucidated by FT-IR, (1)H-NMR and Mass spectra. All newly synthesized compounds were evaluated for their antimycobacterial activities against Mycobacterium tuberculosis H37Rv using the microplatealamar blue assay (MABA). Compounds 4f (5-Nitro-2-furyl analogue) and 4g (5-Nitro-2-thienyl analogue) showed antimycobacterial activity with IC90, 7.57 and 2.96 μg/mL, respectively.