Merck
  • Genotypic analyses of shiga toxin-producing Escherichia coli O157 and non-O157 recovered from feces of domestic animals on rural farms in Mexico.
Genotypic analyses of shiga toxin-producing Escherichia coli O157 and non-O157 recovered from feces of domestic animals on rural farms in Mexico.
PloS one (2012-12-20)
Bianca A Amézquita-López, Beatriz Quiñones, Michael B Cooley, Josefina León-Félix, Nohelia Castro-del Campo, Robert E Mandrell, Maribel Jiménez, Cristóbal Chaidez
ABSTRACT
Shiga toxin-producing Escherichia coli (STEC) are zoonotic enteric pathogens associated with human gastroenteritis worldwide. Cattle and small ruminants are important animal reservoirs of STEC. The present study investigated animal reservoirs for STEC in small rural farms in the Culiacan Valley, an important agricultural region located in Northwest Mexico. A total of 240 fecal samples from domestic animals were collected from five sampling sites in the Culiacan Valley and were subjected to an enrichment protocol followed by either direct plating or immunomagnetic separation before plating on selective media. Serotype O157:H7 isolates with the virulence genes stx2, eae, and ehxA were identified in 40% (26/65) of the recovered isolates from cattle, sheep and chicken feces. Pulse-field gel electrophoresis (PFGE) analysis grouped most O157:H7 isolates into two clusters with 98.6% homology. The use of multiple-locus variable-number tandem repeat analysis (MLVA) differentiated isolates that were indistinguishable by PFGE. Analysis of the allelic diversity of MLVA loci suggested that the O157:H7 isolates from this region were highly related. In contrast to O157:H7 isolates, a greater genotypic diversity was observed in the non-O157 isolates, resulting in 23 PFGE types and 14 MLVA types. The relevant non-O157 serotypes O8:H19, O75:H8, O111:H8 and O146:H21 represented 35.4% (23/65) of the recovered isolates. In particular, 18.5% (12/65) of all the isolates were serotype O75:H8, which was the most variable serotype by both PFGE and MLVA. The non-O157 isolates were predominantly recovered from sheep and were identified to harbor either one or two stx genes. Most non-O157 isolates were ehxA-positive (86.5%, 32/37) but only 10.8% (4/37) harbored eae. These findings indicate that zoonotic STEC with genotypes associated with human illness are present in animals on small farms within rural communities in the Culiacan Valley and emphasize the need for the development of control measures to decrease risks associated with zoonotic STEC.