Merck
  • Prognostic impact of the expression/phosphorylation of the BH3-only proteins of the BCL-2 family in glioblastoma multiforme.
Prognostic impact of the expression/phosphorylation of the BH3-only proteins of the BCL-2 family in glioblastoma multiforme.
Cell death & disease (2012-11-16)
P F Cartron, D Loussouarn, M Campone, S A Martin, F M Vallette
ABSTRACT
Apoptosis has a crucial role in anti-cancer treatment. The proteins of the BCL-2 family are core members of the apoptotic program. Thus, we postulated that alterations in the expression of BCL-2 protein family, and in particular in that of the Bcl-2 homology domain 3 (BH3)-only proteins (which can neutralized anti-apoptotic proteins or activate pro-apoptotic proteins) could account for differences in the overall survival (OS) of patients. To test this hypothesis, we analyzed the expression of 15 members of the BCL-2 protein family (Bax, Bak, Bok, Bcl-2, Bcl-xl, Bcl-w, Mcl-1, Bad, Bid, Bim, Bik, Bmf, Hrk, Noxa and Puma) in glioblastoma multiforme (GBM) tumors, the most frequent brain tumor in adults. We found that none of the individual expression of these proteins is associated with a significant variation in OS of the patients. However, when all BH3 proteins were pooled to determine a BH3(score), this score was significantly correlated with OS of GBM patients. We also noted that patients with a have high level of phospho-Bad and phospho-Bim displayed a lower OS. Thus, BH3 scoring/profiling could be used as an independent prognostic factor in GBM when globally analyzed.