Merck
  • Extracellular vesicle sorting of α-Synuclein is regulated by sumoylation.
Extracellular vesicle sorting of α-Synuclein is regulated by sumoylation.
Acta neuropathologica (2015-03-18)
Marcel Kunadt, Katrin Eckermann, Anne Stuendl, Jing Gong, Belisa Russo, Katrin Strauss, Surya Rai, Sebastian Kügler, Lisandro Falomir Lockhart, Martin Schwalbe, Petranka Krumova, Luis M A Oliveira, Mathias Bähr, Wiebke Möbius, Johannes Levin, Armin Giese, Niels Kruse, Brit Mollenhauer, Ruth Geiss-Friedlander, Albert C Ludolph, Axel Freischmidt, Marisa S Feiler, Karin M Danzer, Markus Zweckstetter, Thomas M Jovin, Mikael Simons, Jochen H Weishaupt, Anja Schneider
ABSTRACT
Extracellular α-Synuclein has been implicated in interneuronal propagation of disease pathology in Parkinson's Disease. How α-Synuclein is released into the extracellular space is still unclear. Here, we show that α-Synuclein is present in extracellular vesicles in the central nervous system. We find that sorting of α-Synuclein in extracellular vesicles is regulated by sumoylation and that sumoylation acts as a sorting factor for targeting of both, cytosolic and transmembrane proteins, to extracellular vesicles. We provide evidence that the SUMO-dependent sorting utilizes the endosomal sorting complex required for transport (ESCRT) by interaction with phosphoinositols. Ubiquitination of cargo proteins is so far the only known determinant for ESCRT-dependent sorting into the extracellular vesicle pathway. Our study reveals a function of SUMO protein modification as a Ubiquitin-independent ESCRT sorting signal, regulating the extracellular vesicle release of α-Synuclein. We deciphered in detail the molecular mechanism which directs α-Synuclein into extracellular vesicles which is of highest relevance for the understanding of Parkinson's disease pathogenesis and progression at the molecular level. We furthermore propose that sumo-dependent sorting constitutes a mechanism with more general implications for cell biology.