Merck
  • Screening for Small-Molecule Modulators of Long Noncoding RNA-Protein Interactions Using AlphaScreen.
Screening for Small-Molecule Modulators of Long Noncoding RNA-Protein Interactions Using AlphaScreen.
Journal of biomolecular screening (2015-07-16)
Roya Pedram Fatemi, Sultan Salah-Uddin, Farzaneh Modarresi, Nathalie Khoury, Claes Wahlestedt, Mohammad Ali Faghihi
ABSTRACT
Long non-protein coding RNAs (lncRNAs) are an important class of molecules that help orchestrate key cellular events. Although their functional roles in cells are not well understood, thousands of lncRNAs and a number of possible mechanisms by which they act have been reported. LncRNAs can exert their regulatory function in cells by interacting with epigenetic enzymes. In this study, we developed a tool to study lncRNA-protein interactions for high-throughput screening of small-molecule modulators using AlphaScreen technology. We tested the interaction of two lncRNAs: brain-derived neurotrophic factor antisense (BDNF-AS) and Hox transcript antisense RNA (HOTAIR), with Enhancer of zeste homolog 2 (EZH2), a histone methyltransferase against a phytochemical library, to look for small-molecule inhibitors that can alter the expression of downstream target genes. We identified ellipticine, a compound that up-regulates BDNF transcription. Our study shows the feasibility of using high-throughput screening to identify modulators of lncRNA-protein interactions and paves the road for targeting lncRNAs that are dysregulated in human disorders using small-molecule therapies.