Merck
First diastereoselective chiral synthesis of (-)-securinine.
Organic letters (2004-01-03)
Toshio Honda, Hidenori Namiki, Kyosuke Kaneda, Hirotake Mizutani
ABSTRACT
[reaction: see text] A diastereoselective total synthesis of securinine in optically pure form was achieved by employing ring-closing metathesis of the corresponding dienyne compound as a key step.