- Pif1 family helicases suppress genome instability at G-quadruplex motifs.
Pif1 family helicases suppress genome instability at G-quadruplex motifs.
Nature (2013-05-10)
Katrin Paeschke, Matthew L Bochman, P Daniela Garcia, Petr Cejka, Katherine L Friedman, Stephen C Kowalczykowski, Virginia A Zakian
PMID23657261
ABSTRACT
The Saccharomyces cerevisiae Pif1 helicase is the prototypical member of the Pif1 DNA helicase family, which is conserved from bacteria to humans. Here we show that exceptionally potent G-quadruplex unwinding is conserved among Pif1 helicases. Moreover, Pif1 helicases from organisms separated by more than 3 billion years of evolution suppressed DNA damage at G-quadruplex motifs in yeast. The G-quadruplex-induced damage generated in the absence of Pif1 helicases led to new genetic and epigenetic changes. Furthermore, when expressed in yeast, human PIF1 suppressed both G-quadruplex-associated DNA damage and telomere lengthening.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)