- A lipophilic thioflavin-T derivative for positron emission tomography (PET) imaging of amyloid in brain.
A lipophilic thioflavin-T derivative for positron emission tomography (PET) imaging of amyloid in brain.
Bioorganic & medicinal chemistry letters (2002-01-30)
Chester A Mathis, Brian J Bacskai, Stephen T Kajdasz, Megan E McLellan, Matthew P Frosch, Bradley T Hyman, Daniel P Holt, Yanming Wang, Guo-Feng Huang, Manik L Debnath, William E Klunk
PMID11814781
ABSTRACT
The synthesis of a new lipophilic thioflavin-T analogue (2-[4' -(methylamino)phenyl]benzothiazole, 6) with high affinity for amyloid is reported. Intravenous injection of [(11)C]-labeled 6 in control mice resulted in high brain uptake. Amyloid deposits were imaged with multiphoton microscopy in the brains of living transgenic mice following the systemic injection of unlabeled 6. [(11)C]6 is a promising amyloid imaging agent for Alzheimer's disease.