Merck
  • Asymmetric Synthesis of alpha-Amino Phosphonic Acids by Diastereoselective Addition of Trimethyl Phosphite onto Chiral Oxazolidines(1).
Asymmetric Synthesis of alpha-Amino Phosphonic Acids by Diastereoselective Addition of Trimethyl Phosphite onto Chiral Oxazolidines(1).
The Journal of organic chemistry (1996-05-31)
Catherine Maury, Tawfik Gharbaoui, Jacques Royer, Henri-Philippe Husson
ABSTRACT
A simple and general asymmetric synthesis of alpha-amino phosphonic acids is described. The method involves the highly selective addition of trialkyl phosphite onto various chiral oxazolidines. Oxazaphosphorinanes thus obtained with an excellent diastereoselectivity furnish the corresponding (S)-alpha-substituted amino phosphonic acids in good overall yields and high ee (77-->97%) after simple deprotection.