- Diametrically disubstituted cyclam derivative having Hg2+-selective fluoroionophoric behaviors.
Diametrically disubstituted cyclam derivative having Hg2+-selective fluoroionophoric behaviors.
The Journal of organic chemistry (2005-03-12)
So-Youn Moon, Na Jin Youn, Sang Mi Park, Suk-Kyu Chang
PMID15760241
ABSTRACT
[reaction: see text] A new fluoroionophore has been synthesized by appending two signaling pyrenylacetamide subunits on the binding motif of 1,8-dimethylcyclam. The designed compound exhibited highly selective and sensitive fluoroionophoric behavior toward Hg(2+) ions of excimer emission in aqueous dioxane (dioxane/H(2)O = 1:9, v/v) solution with a detection limit of 1.3 x 10(-)(6) M. The "ON-OFF" type signaling behavior of the fluoroionophore is due to the metal ion induced conformational changes from folded to open-winged conformations by exploiting the two nearby appended pyrenyl fluorophores.