Merck
Resveratrol Represses Pokemon Expression in Human Glioma Cells.
Molecular neurobiology (2015-04-16)
Yutao Yang, Jiajun Cui, Feng Xue, Anne-Marie Overstreet, Yiping Zhan, Dapeng Shan, Hui Li, Hui Li, Yongjun Wang, Mengmeng Zhang, Chunjiang Yu, Zhi-Qing David Xu
ABSTRACT
POK erythroid myeloid ontogenic factor (Pokemon), an important proto-oncoprotein, is a transcriptional repressor that regulates the expression of many genes and plays an important role in tumorigenesis. Resveratrol (RSV), a natural polyphenolic compound, has many beneficial biological effects on health. In this study, we investigated the role of Pokemon in RSV-induced biological effects and the effect of RSV on the expression of Pokemon in glioma cells. We found that overexpression of Pokemon decreased RSV-induced cell apoptosis, senescence, and anti-proliferative effects. Moreover, we showed that RSV could efficiently decrease the activity of the Pokemon promoter and the expression of Pokemon. Meanwhile, RSV also inhibited Sp1 DNA binding activity to the Pokemon promoter; whereas, it did not influence the expression and nuclear translocation of Sp1. In addition, we found that RSV could increase the recruitment of HDAC1, but decreased p300 to the Pokemon promoter. Taken together, all these results extended our understanding on the anti-cancer mechanism of RSV in glioma cells.