Merck
  • Chemical composition, pretreatments and saccharification of Senna siamea (Lam.) H.S. Irwin & Barneby: An efficient biomass producing tree legume.
Chemical composition, pretreatments and saccharification of Senna siamea (Lam.) H.S. Irwin & Barneby: An efficient biomass producing tree legume.
Bioresource technology (2016-02-19)
Nitesh K Mund, Debabrata Dash, Chitta R Barik, Vaibhav V Goud, Lingaraj Sahoo, Prasannajit Mishra, Nihar R Nayak
ABSTRACT
Protocols were developed for efficient release of glucose from the biomass of Senna siamea, one of the highly efficient biomass producing tree legumes. Composition of mature, 1year and 2years coppice biomass were analysed. For the hydrolysis of the glucan, two pretreatments, cellulose solvent- and organic solvent-based lignocellulose fractionation (COSLIF) and alkali (sodium hydroxide) were used; COSLIF (85% phosphoric acid, 45min incubation at 50°C) pretreated mature biomass exhibited best result in which 88.90% glucose released after 72h of incubation with the use of 5 filter paper units (FPU) of cellulase and 10 international units (IU) of β-glucosidase per gram of glucan. Of the biomass of different particle sizes (40-200mesh) used for saccharification, 40-60mesh shown the maximum glucose release. COSLIF pretreated mature, 1year and 2years coppice biomass showed equivalent glucose release profiles.