Merck
  • Lepidopteran defence droplets - a composite physical and chemical weapon against potential predators.
Lepidopteran defence droplets - a composite physical and chemical weapon against potential predators.
Scientific reports (2016-03-05)
Stefan Pentzold, Mika Zagrobelny, Bekzod Khakimov, Søren Balling Engelsen, Henrik Clausen, Bent Larsen Petersen, Jonas Borch, Birger Lindberg Møller, Søren Bak
ABSTRACT
Insects often release noxious substances for their defence. Larvae of Zygaena filipendulae (Lepidoptera) secrete viscous and cyanogenic glucoside-containing droplets, whose effectiveness was associated with their physical and chemical properties. The droplets glued mandibles and legs of potential predators together and immobilised them. Droplets were characterised by a matrix of an aqueous solution of glycine-rich peptides (H-WG11-NH2) with significant amounts of proteins and glucose. Among the proteins, defensive proteins such as protease inhibitors, proteases and oxidases were abundant. The neurotoxin β-cyanoalanine was also found in the droplets. Despite the presence of cyanogenic glucosides, which release toxic hydrogen cyanide after hydrolysis by a specific β-glucosidase, the only β-glucosidase identified in the droplets (ZfBGD1) was inactive against cyanogenic glucosides. Accordingly, droplets did not release hydrogen cyanide, unless they were mixed with specific β-glucosidases present in the Zygaena haemolymph. Droplets secreted onto the cuticle hardened and formed sharp crystalline-like precipitates that may act as mandible abrasives to chewing predators. Hardening followed water evaporation and formation of antiparallel β-sheets of the peptide oligomers. Consequently, after mild irritation, Zygaena larvae deter predators by viscous and hardening droplets that contain defence proteins and β-cyanoalanine. After severe injury, droplets may mix with exuding haemolymph to release hydrogen cyanide.