Merck
  • Carbon nanotube intramolecular p-i-n junction diodes with symmetric and asymmetric contacts.
Carbon nanotube intramolecular p-i-n junction diodes with symmetric and asymmetric contacts.
Scientific reports (2016-02-27)
Changxin Chen, Chenghao Liao, Liangming Wei, Hanqing Zhong, Rong He, Qinran Liu, Xiaodong Liu, Yunfeng Lai, Chuanjuan Song, Tiening Jin, Yafei Zhang
ABSTRACT
A p-i-n junction diode based on the selectively doped single-walled carbon nanotube (SWCNT) had been investigated, in which two opposite ends of individual SWCNT channel were doped into the p- and n-type SWCNT respectively while the middle segment of SWCNT was kept as the intrinsic. The symmetric and asymmetric contacts were used to fabricate the p-i-n junction diodes respectively and studied the effect of the contact on the device characteristics. It was shown that a low reverse saturation current of ~20 pA could be achieved by these both diodes. We found that the use of the asymmetric contact can effectively improve the performance of the p-i-n diode, with the rectification ratio enhanced from ~10(2) for the device with the Au/Au symmetric contact to >10(3) for the one with the Pd/Al asymmetric contact. The improvement of the device performance by the asymmetric-contact structure was attributed to the decrease of the effective Schottky-barrier height at the contacts under forward bias, increasing the forward current of the diode. The p-i-n diode with asymmetric contact also had a higher rectification ratio than its counterpart before doping the SWCNT channel, which is because that the p-i-n junction in the device decreased the reverse saturated current.