Merck
  • Preparation and evaluation of amoxicillin loaded dual molecularly imprinted nanoparticles for anti-Helicobacter pylori therapy.
Preparation and evaluation of amoxicillin loaded dual molecularly imprinted nanoparticles for anti-Helicobacter pylori therapy.
International journal of pharmaceutics (2015-11-03)
Zhihui Wu, Jiapeng Hou, Yuyan Wang, Miaolin Chai, Yan Xiong, Weiyue Lu, Jun Pan
ABSTRACT
This paper reports studies on preparation and evaluation of amoxicillin loaded dual molecularly imprinted nanoparticles (Amo/Dual-MIPs) designed for anti-H. pylori therapy. Both MNQA and AmoNa were chosen as templates to prepare Dual-MIPs using inverse microemulsion polymerization method. NQA was modified with myristic acid (MNQA) to become amphiphilic and assist in leaving NQA cavities on the surface of Dual-MIPs for H. pylori adhesion. AmoNa was applied to produce imprinting sites in Dual-MIPs for rebinding AmoNa to exert its anti-H. pylori effect. Batch rebinding test demonstrated a preferential rebinding effect of NQA toward the Dual-MIPs. In vivofluorescence imaging showed the prolonged residence time of Dual-MIPs in H. pylori infected mice stomachs after intragastric administration of nanoparticles.In vivo H. pylori clearance tests indicated Amo/Dual-MIPs had a better aniti-H. pylori effect than amoxicillin powder did. In conclusion, Amo/Dual-MIPs may provide an alternative drug delivery strategy for anti-H. pylori therapy.