Merck
  • Porous AgPd-Pd Composite Nanotubes as Highly Efficient Electrocatalysts for Lithium-Oxygen Batteries.
Porous AgPd-Pd Composite Nanotubes as Highly Efficient Electrocatalysts for Lithium-Oxygen Batteries.
Advanced materials (Deerfield Beach, Fla.) (2015-09-25)
Wen-Bin Luo, Xuan-Wen Gao, Shu-Lei Chou, Jia-Zhao Wang, Hua-Kun Liu
ABSTRACT
Porous AgPd-Pd composite nanotubes (NTs) are used as an efficient bifunctional catalyst for the oxygen reduction and evolution reactions in lithium-oxygen batteries. The porous NT structure can facilitate rapid O2 and electrolyte diffusion through the NTs and provide abundant catalytic sites, forming a continuous conductive network throughout the entire energy conversion process, with excellent cycling performance.