Merck
  • Impact of starch content on protein adsorption characteristics in amphiphilic hybrid graft copolymers.
Impact of starch content on protein adsorption characteristics in amphiphilic hybrid graft copolymers.
International journal of biological macromolecules (2015-10-18)
Arijit Sengupta, Allison R Linehan, Peter M Iovine
ABSTRACT
Amphiphilic hybrid graft copolymers were synthesized using a graft-to methodology and their protein adsorption profiles studied. Three different hydrophilic side chains were studied: hydroxypropylated high amylose starch, maltodextrin, and polyethylene glycol (PEG). In the high amylose starch compositions, there was a pronounced decrease in protein adsorption with increasing polysaccharide content. As the starch content in the graft copolymers increased from 10 wt% to 53 wt%, BSA protein adsorption decreased by 83% whereas fibrinogen adsorption was reduced by 40%. Comparisons between the starch-containing hybrid polymers and their respective hydrophobic urethane-linked polyesters were also made. Hybrid 53, containing 53 wt% starch, showed a 85% reduction in BSA adsorption and 51% reduction in fibrinogen relative to their urethane-linked polyester backbone controls. Grafting branched high amylopectin-derived maltodextrin to the synthetic polymer backbones also conferred modest protein resistance to the hydrophobic backbone polymer. Lastly, it was found that a high amylose graft structure provided comparable, if not slightly more effective, protein resistance compared to a similarly constructed PEG-containing amphiphilic copolymer.